Ethyl-2-phenyl-3-(trifluoromethyl)pyrazole-4-carboxylate

Lieferant: Apollo Scientific
Achtung

PC31628-1G
APOSPC31628-1GEA 59.6 EUR
APOSPC31628-1G
Ethyl-2-phenyl-3-(trifluoromethyl)pyrazole-4-carboxylate
Ethyl-2-phenyl-3-(trifluoromethyl)pyrazole-4-carboxylate
Formel: C₁₃H₁₁F₃N₂O₂
Molecular Weight: 284,24 g/mol
Siedepunkt: 125 °C/0,06mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00068138
CAS-Nummer: 112055-34-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR