3-Brom-2-fluorbenzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Bromo-2-fluorophenyl)methanol, 3-Bromo-2-fluorobenzyl alcohol, 1-Bromo-2-fluoro-3-(hydroxymethyl)benzene

PC3167-1G
APOSPC3167-1GEA 21.9 EUR
APOSPC3167-1G
3-Brom-2-fluorbenzylalkohol 98%
3-Brom-2-fluorbenzylalkohol
Formel: C₇H₆BrFO
Molecular Weight: 205,0200 g/mol
Siedepunkt: 104…108 °C/4mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD09842441
CAS-Nummer: 261723-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR