3-Amino-4-fluorbenzylalkohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Amino-4-fluorophenyl)methanol

PC3172-500MG
APOSPC3172-500MGEA 473 EUR
APOSPC3172-500MG
3-Amino-4-fluorbenzylalkohol
3-Amino-4-fluorbenzylalkohol
Formel: C₇H₈FNO
Molecular Weight: 141,1429 g/mol
Schmelzpunkt: 76…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09865016
CAS-Nummer: 227609-86-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR