4'-Chlor-2,2,2-trifluoracetophenon 97%

Lieferant: Apollo Scientific

Synonyme: 1-(4-Chlorophenyl)-2,2,2-trifluoroethanone

PC31735-5G
APOSPC31735-5GEA 75.4 EUR
APOSPC31735-5G
4'-Chlor-2,2,2-trifluoracetophenon 97%
4'-Chlor-2,2,2-trifluoracetophenon
Formel: C₈H₄ClF₃O
Molecular Weight: 208,5700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00018686
CAS-Nummer: 321-37-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR