3-(3-Fluoro-4-methylphenyl)-1,2,4-thiadiazol-5-ylamine

Lieferant: Apollo Scientific

Synonyme: 5-Amino-3-(3-fluoro-4-methylphenyl)-1,2,4-thiadiazole

PC3175-500MG
APOSPC3175-500MGEA 434 EUR
APOSPC3175-500MG
3-(3-Fluoro-4-methylphenyl)-1,2,4-thiadiazol-5-ylamine
3-(3-Fluoro-4-methylphenyl)-1,2,4-thiadiazol-5-ylamine
Formel: C₉H₈FN₃S
Molecular Weight: 209,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09865007
CAS-Nummer: 1017782-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR