8-Bromo-4-chloro-2-(trifluoromethyl)quinoline

Lieferant: Apollo Scientific
Achtung

PC31854-250MG PC31854-1G PC31854-5G
APOSPC31854-250MGEA 0 EUR
APOSPC31854-250MG APOSPC31854-1G APOSPC31854-5G
8-Bromo-4-chloro-2-(trifluoromethyl)quinoline
8-Bromo-4-chloro-2-(trifluoromethyl)quinoline
Formel: C₁₀H₄BrClF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD00174293
CAS-Nummer: 655235-61-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR