Methyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylate, 3-Chloro-2-(methoxycarbonyl)-5-(trifluoromethyl)pyridine, Methyl 3-chloro-5-(trifluoromethyl)picolinate

PC31856-250MG
APOSPC31856-250MGEA 21.9 EUR
APOSPC31856-250MG
Methyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate
Methyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate
Formel: C₈H₅ClF₃NO₂
Molecular Weight: 239,579 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01570523
CAS-Nummer: 655235-65-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR