4-Chloro-6-(trifluoromethoxy)-2-(trifluoromethyl)quinoline

Lieferant: Apollo Scientific

PC31863-1G
APOSPC31863-1GEA 183 EUR
APOSPC31863-1G
4-Chloro-6-(trifluoromethoxy)-2-(trifluoromethyl)quinoline
4-Chloro-6-(trifluoromethoxy)-2-(trifluoromethyl)quinoline
Formel: C₁₁H₄ClF₆NO
Molecular Weight: 315,599 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01566524
CAS-Nummer: 503148-24-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR