3-(2-Chloro-6-fluorophenyl)-N-(2-{[(fur-2-yl)methyl]sulphanyl}ethyl)-5-methylisoxazole-4-carboxamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(2-Chloro-6-fluorophenyl)-N-(2-{[(fur-2-yl)methyl]thio}ethyl)-5-methylisoxazole-4-carboxamide, 3-(2-Chloro-6-fluorophenyl)-N-(2-{[(fur-2-yl)methyl]sulphanyl}ethyl)-5-methyl-1,2-oxazole-4-carboxamide

PC31870-250MG
APOSPC31870-250MGEA 71.2 EUR
APOSPC31870-250MG
3-(2-Chloro-6-fluorophenyl)-N-(2-{[(fur-2-yl)methyl]sulphanyl}ethyl)-5-methylisoxazole-4-carboxamide
3-(2-Chloro-6-fluorophenyl)-N-(2-{[(fur-2-yl)methyl]sulphanyl}ethyl)-5-methylisoxazole-4-carboxamide
Formel: C₁₈H₁₆ClFN₂O₃S
Molecular Weight: 394,848 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01566569
CAS-Nummer: 680211-94-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR