4-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid

Lieferant: Apollo Scientific
Achtung

PC31875-1G
APOSPC31875-1GEA 116 EUR
APOSPC31875-1G
4-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid
4-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid
Formel: C₁₁H₇F₃N₂O₂
Schmelzpunkt: 189…190 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03086203
CAS-Nummer: 220462-27-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR