1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Fluorophenyl)-4-formyl-5-methyl-1H-pyrazole

PC31881-1G
APOSPC31881-1GEA 193 EUR
APOSPC31881-1G
1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carboxaldehyde
1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carboxaldehyde
Formel: C₁₁H₉FN₂O
Molecular Weight: 204,2 g/mol
Schmelzpunkt: 111…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03407349
CAS-Nummer: 423768-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR