5-Amino-4-[4-(trifluoromethyl)phenyl]-1,2,3-thiadiazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(5-Amino-1,2,3-thiadiazol-4-yl)benzotrifluoride, 4-[4-(Trifluoromethyl)phenyl]-1,2,3-thiadiazol-5-amine

PC31885-250MG
APOSPC31885-250MGEA 174 EUR
APOSPC31885-250MG
5-Amino-4-[4-(trifluoromethyl)phenyl]-1,2,3-thiadiazole
5-Amino-4-[4-(trifluoromethyl)phenyl]-1,2,3-thiadiazole
Formel: C₉H₆F₃N₃S
Molecular Weight: 245,224 g/mol
Schmelzpunkt: 125…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03407371
CAS-Nummer: 423769-76-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR