4-Chlor-3-(trifluormethyl)benzolsulfonylchlorid 96%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Chlor-3-(trifluormethyl)benzol-1-sulfonylchlorid, 4-Chlor-3-(trifluormethyl)benzolsulfonsäurechlorid, 4-Chlor-3-(trifluormethyl)benzolsulfochlorid

PC31887-1G
APOSPC31887-1GEA 14.7 EUR
APOSPC31887-1G
4-Chlor-3-(trifluormethyl)benzolsulfonylchlorid 96%
4-Chlor-3-(trifluormethyl)benzolsulfonylchlorid
Formel: C₇H₃Cl₂F₃O₂S
Molecular Weight: 279,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00024882
CAS-Nummer: 32333-53-2
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR