1-(4-Fluorophenyl)-3-(4-hydroxyphenyl)-2-propen-1-one
Lieferant: Apollo Scientific
PC31955-1G
APOSPC31955-1GEA
161
EUR
APOSPC31955-1G
1-(4-Fluorophenyl)-3-(4-hydroxyphenyl)-2-propen-1-one
1-(4-Fluorophenyl)-3-(4-hydroxyphenyl)-2-propen-1-one
Formel:
C₁₅H₁₁FO₂ Schmelzpunkt: 189…190 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00206942 CAS-Nummer: 7397-22-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...