5-[3,5-bis(trifluoromethyl)phenyl]-3-(chloromethyl)-1,2,4-oxadiazole
Lieferant: Apollo Scientific
|
Gefahr
|
PC32017-1G
APOSPC32017-1GEA
59.6
EUR
APOSPC32017-1G
5-[3,5-bis(trifluoromethyl)phenyl]-3-(chloromethyl)-1,2,4-oxadiazole
5-[3,5-bis(trifluoromethyl)phenyl]-3-(chloromethyl)-1,2,4-oxadiazole
Formel:
C₁₁H₅ClF₆N₂O Molecular Weight: 330,614 g/mol Schmelzpunkt: 58…59 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01764514 CAS-Nummer: 287198-14-5 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...