N-Ethyl-2-fluoroaniline 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC3207-10G
PC3207-1G
PC3207-5G
APOSPC3207-10GEA
0
EUR
APOSPC3207-10G
APOSPC3207-1G
APOSPC3207-5G
N-Ethyl-2-fluoroaniline 95%
N-Ethyl-2-fluoroaniline
Formel:
C₈H₁₀FN Siedepunkt: 72…74 °C (15 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00203991 CAS-Nummer: 2707-64-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...