4-(Trifluormethoxy)anilin Hydrochlorid

Lieferant: Apollo Scientific
Achtung

PC32088-1G PC32088-5G
APOSPC32088-1GEA 0 EUR
APOSPC32088-1G APOSPC32088-5G
4-(Trifluormethoxy)anilin Hydrochlorid
4-(Trifluormethoxy)anilin Hydrochlorid
Formel: C₇H₇ClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00219956
CAS-Nummer: 42823-24-5
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR