6-Chloro-3-[4-(trifluoromethyl)phenyl][1,2,4]triazolo[4,3-b]pyridazine

Lieferant: Apollo Scientific

PC32092-500MG
APOSPC32092-500MGEA 183 EUR
APOSPC32092-500MG
6-Chloro-3-[4-(trifluoromethyl)phenyl][1,2,4]triazolo[4,3-b]pyridazine
6-Chloro-3-[4-(trifluoromethyl)phenyl][1,2,4]triazolo[4,3-b]pyridazine
Formel: C₁₂H₆ClF₃N₄
Molecular Weight: 298,651 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00277273
CAS-Nummer: 175204-95-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR