2'-Fluor-5'-nitroacetophenon

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Fluoro-5-nitrophenyl)ethanone

PC32093-25G PC32093-5G PC32093-100G
APOSPC32093-25GEA 59.6 EUR
APOSPC32093-25G APOSPC32093-5G APOSPC32093-100G
2'-Fluor-5'-nitroacetophenon
2'-Fluor-5'-nitroacetophenon
Formel: C₈H₆FNO₃
Molecular Weight: 183.1400 g/mol
Schmelzpunkt: 46…50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00277867
CAS-Nummer: 79110-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR