N-(2,6-Difluoro-3-nitrophenyl)acetamide 95+%

Lieferant: Apollo Scientific

PC32095-1G
APOSPC32095-1GEA 132 EUR
APOSPC32095-1G
N-(2,6-Difluoro-3-nitrophenyl)acetamide 95+%
N-(2,6-Difluoro-3-nitrophenyl)acetamide
Formel: C₈H₆F₂N₂O₃
Molecular Weight: 216,1400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00174326
CAS-Nummer: 25892-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR