Ethyl 1-(2,4-difluorophenyl)-5-methylpyrazole-4-carboxylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC3211-1G
APOSPC3211-1GEA
434
EUR
APOSPC3211-1G
Ethyl 1-(2,4-difluorophenyl)-5-methylpyrazole-4-carboxylate 95%
Ethyl 1-(2,4-difluorophenyl)-5-methylpyrazole-4-carboxylate
Formel:
C₁₃H₁₂F₂N₂O₂ Molecular Weight: 266,243 g/mol Siedepunkt: 116…118 °C (0,5 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067830 CAS-Nummer: 175135-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...