3-Brom-4-(trifluormethoxy)anilin 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC3215-100G
PC3215-25G
PC3215-5G
APOSPC3215-100GEA
0
EUR
APOSPC3215-100G
APOSPC3215-25G
APOSPC3215-5G
3-Brom-4-(trifluormethoxy)anilin 98%
3-Brom-4-(trifluormethoxy)anilin
Formel:
C₇H₅BrF₃NO Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00190127 CAS-Nummer: 191602-54-7 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...