Ethyl-4-hydroxy-8-(trifluormethoxy)chinolin-3-carboxylat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 4-hydroxy-8-(trifluoromethoxy)quinoline-3-carboxylate

PC3216-1G
APOSPC3216-1GEA 29 EUR
APOSPC3216-1G
Ethyl-4-hydroxy-8-(trifluormethoxy)chinolin-3-carboxylat 97%
Ethyl-4-hydroxy-8-(trifluormethoxy)chinolin-3-carboxylat
Formel: C₁₃H₁₀F₃NO₄
Molecular Weight: 301,2180 g/mol
Schmelzpunkt: 235…240 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00221480
CAS-Nummer: 175278-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR