3-Amino-4-fluorophenylboronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Borono-2-fluoroaniline, Boronic acid, B-(3-amino-4-fluorophenyl)-, 3-Amino-4-fluorobenzeneboronic acid
PC3218-1G
APOSPC3218-1GEA
183
EUR
APOSPC3218-1G
3-Amino-4-fluorophenylboronic acid
3-Amino-4-fluorophenylboronic acid
Formel:
C₆H₇BFNO₂ Molecular Weight: 154,94 g/mol Schmelzpunkt: 226…232 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03092925 CAS-Nummer: 873566-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...