4-Cyan-2-fluorphenylboronsäurepinakolester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Cyano-2-fluorobenzeneboronic acid, pinacol ester, 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 4-Cyano-2-fluorobenzeneboronic acid pinacol ester

PC3223-1G PC3223-250MG
APOSPC3223-1GEA 289 EUR
APOSPC3223-1G APOSPC3223-250MG
4-Cyan-2-fluorphenylboronsäurepinakolester 98%
4-Cyan-2-fluorphenylboronsäurepinakolester
Formel: C₁₃H₁₅BFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09998162
CAS-Nummer: 1035235-29-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR