4-Amino-N-(2,4-difluorphenyl)benzolsulfonamid
Lieferant: Apollo Scientific
|
Achtung
|
PC32230-250MG
APOSPC32230-250MGEA
68.3
EUR
APOSPC32230-250MG
4-Amino-N-(2,4-difluorphenyl)benzolsulfonamid
4-Amino-N-(2,4-difluorphenyl)benzolsulfonamid
Formel:
C₁₂H₁₀F₂N₂O₂S Schmelzpunkt: 161…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01935782 CAS-Nummer: 1717-36-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...