5-Oxo-1-[3-(trifluoromethyl)phenyl]pyrrolidine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC3224-500MG
APOSPC3224-500MGEA 109 EUR
APOSPC3224-500MG
5-Oxo-1-[3-(trifluoromethyl)phenyl]pyrrolidine-3-carboxylic acid
5-Oxo-1-[3-(trifluoromethyl)phenyl]pyrrolidine-3-carboxylic acid
Formel: C₁₂H₁₀F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00665765
CAS-Nummer: 2357-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR