Pentafluorophenyl 1-(thien-2-ylmethyl)piperidine-4-carboxylate 97%
Lieferant: Apollo Scientific
PC3225-250MG
APOSPC3225-250MGEA
364
EUR
APOSPC3225-250MG
Pentafluorophenyl 1-(thien-2-ylmethyl)piperidine-4-carboxylate 97%
Pentafluorophenyl 1-(thien-2-ylmethyl)piperidine-4-carboxylate
Formel:
C₁₇H₁₄F₅NO₂S Molecular Weight: 391,356 g/mol Schmelzpunkt: 73…75 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09879954 CAS-Nummer: 930111-06-1 UN: 2928 ADR: 6.1+8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...