4-Amino-N-[4-(trifluoromethyl)phenyl]benzenesulphonamide

Lieferant: Apollo Scientific
Achtung

PC32257-250MG
APOSPC32257-250MGEA 0 EUR
APOSPC32257-250MG
4-Amino-N-[4-(trifluoromethyl)phenyl]benzenesulphonamide
4-Amino-N-[4-(trifluoromethyl)phenyl]benzenesulphonamide
Formel: C₁₃H₁₁F₃N₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD01935804
CAS-Nummer: 339-42-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR