(2E)-1-(Fur-2-yl)-3-[4-(trifluoromethyl)phenyl]prop-2-en-1-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-[(1E)-3-(Fur-2-yl)-3-oxoprop-1-en-1-yl]benzotrifluoride, 2-{(2E)-3-[4-(Trifluoromethyl)phenyl]prop-2-enoyl}furan
PC32266-250MG
APOSPC32266-250MGEA
360
EUR
APOSPC32266-250MG
(2E)-1-(Fur-2-yl)-3-[4-(trifluoromethyl)phenyl]prop-2-en-1-one
(2E)-1-(Fur-2-yl)-3-[4-(trifluoromethyl)phenyl]prop-2-en-1-one
Formel:
C₁₄H₉F₃O₂ Molecular Weight: 266,215 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00176655 CAS-Nummer: 680215-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...