4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole

Lieferant: Apollo Scientific
Gefahr

PC3229-1G
APOSPC3229-1GEA 179 EUR
APOSPC3229-1G
4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole
4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole
Formel: C₁₂H₁₀BrF₃N₂O
Molecular Weight: 335,12 g/mol
Schmelzpunkt: 75,5…77.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09879947
CAS-Nummer: 912569-72-3
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR