3-Amino-6-methyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carbonitrile

Lieferant: Apollo Scientific

PC32294-1G
APOSPC32294-1GEA 125 EUR
APOSPC32294-1G
3-Amino-6-methyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carbonitrile
3-Amino-6-methyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carbonitrile
Formel: C₁₀H₆F₃N₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00486825
CAS-Nummer: 317840-08-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR