3-Amino-6-phenyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC32295-1G
APOSPC32295-1GEA
174
EUR
APOSPC32295-1G
3-Amino-6-phenyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxylic acid
3-Amino-6-phenyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxylic acid
Formel:
C₁₅H₉F₃N₂O₂S Molecular Weight: 338,304 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00398797 CAS-Nummer: 104960-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...