5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(4-(Trifluoromethyl)phenyl)-1H-tetrazole
PC32296-1G
APOSPC32296-1GEA
87
EUR
APOSPC32296-1G
5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole
5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole
Formel:
C₈H₅F₃N₄ Molecular Weight: 214,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06248886 CAS-Nummer: 2251-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...