Ethyl-2-(4-fluorophenyl)-3-(trifluoromethyl)pyrazole-4-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

PC3233-250MG
APOSPC3233-250MGEA 85.7 EUR
APOSPC3233-250MG
Ethyl-2-(4-fluorophenyl)-3-(trifluoromethyl)pyrazole-4-carboxylate 95%
Ethyl-2-(4-fluorophenyl)-3-(trifluoromethyl)pyrazole-4-carboxylate
Formel: C₁₃H₁₀F₄N₂O₂
Molecular Weight: 302,23 g/mol
Siedepunkt: 140…142 °C/0,2mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00173869
CAS-Nummer: 175137-38-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR