3-Nitro-1,1,1-trifluoropropan-2-ol 97%
Lieferant: Apollo Scientific
PC32425-25G
APOSPC32425-25GEA
1790
EUR
APOSPC32425-25G
3-Nitro-1,1,1-trifluoropropan-2-ol 97%
3-Nitro-1,1,1-trifluoropropan-2-ol
Formel:
C₃H₄F₃NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01328287 CAS-Nummer: 453-35-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...