1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-1H-pyrrole

Lieferant: Apollo Scientific

PC32453-1G
APOSPC32453-1GEA 277 EUR
APOSPC32453-1G
1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-1H-pyrrole
1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-1H-pyrrole
Formel: C₁₁H₆Cl₂F₃N
Molecular Weight: 280,073 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00278514
CAS-Nummer: 259175-24-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR