5-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-2-furaldehyde

Lieferant: Apollo Scientific

PC32459-1G
APOSPC32459-1GEA 232 EUR
APOSPC32459-1G
5-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-2-furaldehyde
5-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-2-furaldehyde
Formel: C₁₂H₅Cl₂F₃O₂
Molecular Weight: 309,068 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00278526
CAS-Nummer: 680215-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR