2'-Fluorbiphenyl-4-carbonsäure
Lieferant: Apollo Scientific
Synonyme:
2'-Fluoro-[1,1'-biphenyl]-4-carboxylic acid
PC32477-1G
APOSPC32477-1GEA
149
EUR
APOSPC32477-1G
2'-Fluorbiphenyl-4-carbonsäure
2'-Fluorbiphenyl-4-carbonsäure
Formel:
C₁₃H₉FO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00278732 CAS-Nummer: 365-12-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...