4-Hydroxy-3-(trifluormethyl)benzaldehyd

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Formyl-2-(trifluoromethyl)phenol, 5-Formyl-2-hydroxybenzotrifluoride

PC3247Q-100MG
APOSPC3247Q-100MGEA 21.9 EUR
APOSPC3247Q-100MG
4-Hydroxy-3-(trifluormethyl)benzaldehyd
4-Hydroxy-3-(trifluoromethyl)benzaldehyde
Formel: C₈H₅F₃O₂
Schmelzpunkt: 158…165 °C
MDL: MFCD01091006
CAS-Nummer: 220227-98-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR