3-[3-(Trifluoromethoxy)phenyl]isoxazole

Lieferant: Apollo Scientific
Achtung

PC32508-1G PC32508-250MG
APOSPC32508-1GEA 98.9 EUR
APOSPC32508-1G APOSPC32508-250MG
3-[3-(Trifluoromethoxy)phenyl]isoxazole
3-[3-(Trifluoromethoxy)phenyl]isoxazole
Formel: C₁₀H₆F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01313379
CAS-Nummer: 260046-96-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR