Ethyl perfluoro-n-dodecanoate 97%

Lieferant: Apollo Scientific
Achtung

PC3250M-1G
APOSPC3250M-1GEA 29.2 EUR
APOSPC3250M-1G
Ethyl perfluoro-n-dodecanoate 97%
Ethyl perfluoro-n-dodecanoate
Formel: C₁₄H₅F₂₃O₂
Siedepunkt: 96 °C/9mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00236676
CAS-Nummer: 158607-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR