2-Chloro-1-{4-[3-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}ethan-1-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[4-(Chloroacetyl)piperazin-1-yl]-3-(trifluoromethyl)pyridine, 1-(Chloroacetyl)-4-[3-(trifluoromethyl)pyridin-2-yl]piperazine

PC32512-250MG
APOSPC32512-250MGEA 46.6 EUR
APOSPC32512-250MG
2-Chloro-1-{4-[3-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}ethan-1-one
2-Chloro-1-{4-[3-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}ethan-1-one
Formel: C₁₂H₁₃ClF₃N₃O
Molecular Weight: 307,699 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01571119
CAS-Nummer: 260392-48-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR