N-[2-(2,4-Dichlorophenoxy)phenyl]-2,6-difluoro-3-nitrobenzamide

Lieferant: Apollo Scientific
Achtung

PC32533-50MG
APOSPC32533-50MGEA 0 EUR
APOSPC32533-50MG
N-[2-(2,4-Dichlorophenoxy)phenyl]-2,6-difluoro-3-nitrobenzamide
N-[2-(2,4-Dichlorophenoxy)phenyl]-2,6-difluoro-3-nitrobenzamide
Formel: C₁₉H₁₀Cl₂F₂N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD01567171
CAS-Nummer: 260553-38-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR