5-[4-Fluoro-3-(trifluoromethyl)phenyl]-2-furaldehyde

Lieferant: Apollo Scientific

PC32543-1G
APOSPC32543-1GEA 212 EUR
APOSPC32543-1G
5-[4-Fluoro-3-(trifluoromethyl)phenyl]-2-furaldehyde
5-[4-Fluoro-3-(trifluoromethyl)phenyl]-2-furaldehyde
Formel: C₁₂H₆F₄O₂
Molecular Weight: 258,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01571138
CAS-Nummer: 306936-05-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR