Ethyl 5-methyl-2-(trifluoromethyl)-3-furoate 97%

Lieferant: Apollo Scientific

PC3255-1G
APOSPC3255-1GEA 232 EUR
APOSPC3255-1G
Ethyl 5-methyl-2-(trifluoromethyl)-3-furoate 97%
Ethyl 5-methyl-2-(trifluoromethyl)-3-furoate
Formel: C₉H₉F₃O₃
Molecular Weight: 222,161 g/mol
Siedepunkt: 55 °C (0,3 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00275564
CAS-Nummer: 17515-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR