Methyl 3-[4-(trifluoromethoxy)phenyl]acrylate

Lieferant: Apollo Scientific

PC32550-1G
APOSPC32550-1GEA 0 EUR
APOSPC32550-1G
Methyl 3-[4-(trifluoromethoxy)phenyl]acrylate
Methyl 3-[4-(trifluoromethoxy)phenyl]acrylate
Formel: C₁₁H₉F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01571241
CAS-Nummer: 1226460-04-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR