Ethyl 2-{2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetate

Lieferant: Apollo Scientific

PC32561-1G PC32561-5G
APOSPC32561-1GEA 132 EUR
APOSPC32561-1G APOSPC32561-5G
Ethyl 2-{2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetate
Ethyl 2-{2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetate
Formel: C₁₄H₁₂F₃NO₂S
Schmelzpunkt: 37,8…40.6 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01765079
CAS-Nummer: 680215-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR