3-[4-(Trifluoromethyl)phenyl]-1H-pyrazole-4-carboxaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
PC32577-1G
APOSPC32577-1GEA
0
EUR
APOSPC32577-1G
3-[4-(Trifluoromethyl)phenyl]-1H-pyrazole-4-carboxaldehyde
3-[4-(Trifluoromethyl)phenyl]-1H-pyrazole-4-carboxaldehyde
Formel:
C₁₁H₇F₃N₂O Schmelzpunkt: 113…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02677691 CAS-Nummer: 306936-65-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...