Ethyl nonafluorovalerate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl Perfluorovalerate, Nonafluorovaleric Acid Ethyl Ester, Perfluorovaleric Acid Ethyl Ester

PC3260-5G
APOSPC3260-5GEA 21.9 EUR
APOSPC3260-5G
Ethyl nonafluorovalerate 98%
Ethyl nonafluorovalerate
Formel: C₇H₅F₉O₂
Molecular Weight: 292,10 g/mol
Siedepunkt: 121 °C
Dichte: 1,48 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00077523
CAS-Nummer: 424-36-2
UN: 3272
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR